CAS RN 111787-89-4
CAS RN 111787-89-4
IUPAC Name:
5-(4-chlorophenyl)-2-methylfuran-3-carboxylic acid
Synonyms:
5-(4-chlorophenyl)-2-methylfuran-3-carboxylic acid
Molecular Weight (g/mol):
236.65
Molecular Formula:
C12H9ClO3
InChi Key:
HUQOLZUHGMRQFS-UHFFFAOYSA-N
SMILES:
CC1=C(C=C(O1)C1=CC=C(Cl)C=C1)C(O)=O
1
–
1
of
1
results
1
5-(4-Chlorophenyl)-2-methyl-3-furoic acid, ≥97%, Thermo Scientific™
CAS: 111787-89-4 Molecular Formula: C12H9ClO3 Molecular Weight (g/mol): 236.65 MDL Number: MFCD00275560 InChI Key: HUQOLZUHGMRQFS-UHFFFAOYSA-N PubChem CID: 2742181 IUPAC Name: 5-(4-chlorophenyl)-2-methylfuran-3-carboxylic acid SMILES: CC1=C(C=C(O1)C1=CC=C(Cl)C=C1)C(O)=O