CAS RN 112-20-9
CAS RN 112-20-9
IUPAC Name:
nonan-1-aminium chloride
Synonyms:
nonan-1-aminium chloride
Molecular Weight (g/mol):
179.73
Molecular Formula:
C9H22ClN
InChi Key:
IMGHCWMUWVHYCO-UHFFFAOYSA-N
SMILES:
[Cl-].CCCCCCCCC[NH3+]
1
–
2
of
2
results
1
1-Nonylamine, 97%, Thermo Scientific Chemicals
CAS: 112-20-9 Molecular Formula: C9H22ClN Molecular Weight (g/mol): 179.73 MDL Number: MFCD00008249 InChI Key: IMGHCWMUWVHYCO-UHFFFAOYSA-N Synonym: nonylamine,n-nonylamine,1-aminononane,1-nonanamine,1-nonylamine,unii-7l7h2ht4ok,7l7h2ht4ok,nonyl-amine,n-nonyl amine,amine c9 PubChem CID: 16215 IUPAC Name: nonan-1-amine SMILES: [Cl-].CCCCCCCCC[NH3+]
2
n-Nonylamine, 98%, Thermo Scientific Chemicals
CAS: 112-20-9 Molecular Formula: C9H22ClN Molecular Weight (g/mol): 179.73 MDL Number: MFCD00008249 InChI Key: IMGHCWMUWVHYCO-UHFFFAOYSA-N Synonym: nonylamine,n-nonylamine,1-aminononane,1-nonanamine,1-nonylamine,unii-7l7h2ht4ok,7l7h2ht4ok,nonyl-amine,n-nonyl amine,amine c9 PubChem CID: 16215 IUPAC Name: nonan-1-amine SMILES: [Cl-].CCCCCCCCC[NH3+]