man

Test

CAS RN 1121-25-1

CH3NHO

CAS RN 1121-25-1

IUPAC Name: 2-methylpyridin-3-ol
Synonyms: 2-methylpyridin-3-ol
Molecular Weight (g/mol): 109.13
Molecular Formula: C6H7NO
InChi Key: AQSRRZGQRFFFGS-UHFFFAOYSA-N
SMILES: CC1=NC=CC=C1O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results