CAS RN 112245-09-7
CAS RN 112245-09-7
IUPAC Name:
2-amino-3,3-dimethylbutan-1-ol
Synonyms:
2-amino-3,3-dimethylbutan-1-ol
Molecular Weight (g/mol):
117.19
Molecular Formula:
C6H15NO
InChi Key:
JBULSURVMXPBNA-UHFFFAOYNA-N
SMILES:
CC(C)(C)C(N)CO