man

Test

CAS RN 1123-85-9

CH3OH

CAS RN 1123-85-9

IUPAC Name: 2-phenylpropan-1-ol
Synonyms: 2-phenylpropanol-1 hydratropic alcohol
Molecular Weight (g/mol): 136.19
Molecular Formula: C9H12O
InChi Key: RNDNSYIPLPAXAZ-UHFFFAOYNA-N
SMILES: CC(CO)C1=CC=CC=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results