CAS RN 1124-20-5
CAS RN 1124-20-5
IUPAC Name:
1-methyl-3-(prop-1-en-2-yl)benzene
Synonyms:
1-methyl-3-ethenyl benzene
Molecular Weight (g/mol):
132.21
Molecular Formula:
C10H12
InChi Key:
XXTQHVKTTBLFRI-UHFFFAOYSA-N
SMILES:
CC(=C)C1=CC=CC(C)=C1
1
–
1
of
1
results
1
alpha,3-Dimethylstyrene, Thermo Scientific Chemicals
CAS: 1124-20-5 Molecular Formula: C10H12 Molecular Weight (g/mol): 132.206 MDL Number: MFCD08060972 InChI Key: XXTQHVKTTBLFRI-UHFFFAOYSA-N Synonym: m,alpha-dimethylstyrene,alpha,3-dimethylstyrene,m-isopropenyltoluene,unii-3ey8gvm0dx,benzene, 1-methyl-3-1-methylethenyl,3ey8gvm0dx,1-methyl-3-prop-1-en-2-yl benzene,1-isopropenyl-3-methyl-benzene,1-methyl-3-1-methylethenyl benzene,1-methyl-3-ethenyl benzene PubChem CID: 70759 IUPAC Name: 1-methyl-3-prop-1-en-2-ylbenzene SMILES: CC1=CC=CC(=C1)C(=C)C