man

Test

CAS RN 1132-05-4

CH3OOHH2C

CAS RN 1132-05-4

IUPAC Name: 1-[4-hydroxy-3-(prop-2-en-1-yl)phenyl]ethan-1-one
Synonyms: 1-[4-hydroxy-3-(prop-2-en-1-yl)phenyl]ethanone
Molecular Weight (g/mol): 176.22
Molecular Formula: C11H12O2
InChi Key: XVTCWUFLNLZPEJ-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(O)C(CC=C)=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results