man

Test

CAS RN 114344-60-4

BrH2NN

CAS RN 114344-60-4

IUPAC Name: 2-amino-3-bromobenzonitrile
Synonyms: 2-amino-3-bromobenzonitrile
Molecular Weight (g/mol): 197.04
Molecular Formula: C7H5BrN2
InChi Key: RTIWACSVMFUEBF-UHFFFAOYSA-N
SMILES: NC1=C(C=CC=C1Br)C#N

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results