man

Test

CAS RN 114346-31-5

H3CH3CCH3OONHHNOOOHO

CAS RN 114346-31-5

IUPAC Name: 2-{[(tert-butoxy)carbonyl]amino}-3-[4-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)phenyl]propanoic acid
Synonyms: 2-[(tert-butoxycarbonyl)amino]-3-(4-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}phenyl)propanoic acid
Molecular Weight (g/mol): 502.57
Molecular Formula: C29H30N2O6
InChi Key: ZKSJJSOHPQQZHC-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC(CC1=CC=C(NC(=O)OCC2C3=CC=CC=C3C3=CC=CC=C23)C=C1)C(O)=O

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results