man

Test

CAS RN 115-18-4

CH3CH3HOH2C

CAS RN 115-18-4

IUPAC Name: 2-methylbut-3-en-2-ol
Synonyms: 2-methyl-3-buten-2-ol
Molecular Weight (g/mol): 86.13
Molecular Formula: C5H10O
InChi Key: HNVRRHSXBLFLIG-UHFFFAOYSA-N
SMILES: CC(C)(O)C=C
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)