man

Test

CAS RN 115444-73-0

CH3NNOOH

CAS RN 115444-73-0

IUPAC Name: 4-(2-methyl-1H-1,3-benzodiazol-1-yl)butanoic acid
Synonyms: 4-(2-methyl-1,3-benzodiazol-1-yl)butanoic acid
Molecular Weight (g/mol): 218.26
Molecular Formula: C12H14N2O2
InChi Key: IRBRKNDBYUBKIR-UHFFFAOYSA-N
SMILES: CC1=NC2=CC=CC=C2N1CCCC(O)=O

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results