man

Test

CAS RN 115532-50-8

CH3NCH3NCH3H3COOOCH3OClOOO

CAS RN 115532-50-8

IUPAC Name: 6-(dimethylamino)-9-[2-(methoxycarbonyl)phenyl]-N,N-dimethyl-3H-xanthen-3-iminium perchlorate
Synonyms: 6-(dimethylamino)-9-[2-(methoxycarbonyl)phenyl]-N,N-dimethylxanthen-3-iminium perchlorate
Molecular Weight (g/mol): 500.93
Molecular Formula: C25H25ClN2O7
InChi Key: PFYWPQMAWCYNGW-UHFFFAOYSA-M
SMILES: [O-][Cl](=O)(=O)=O.COC(=O)C1=CC=CC=C1C1=C2C=CC(C=C2OC2=CC(=CC=C12)N(C)C)=[N+](C)C

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results