CAS RN 116422-39-0
CAS RN 116422-39-0
IUPAC Name:
(2S)-2-methoxypropan-1-ol
Synonyms:
(2S)-2-methoxypropan-1-ol
Molecular Weight (g/mol):
90.12
Molecular Formula:
C4H10O2
InChi Key:
YTTFFPATQICAQN-BYPYZUCNSA-N
SMILES:
CO[C@@H](C)CO
1
–
1
of
1
results
1
(S)-(+)-2-Methoxypropanol, 97%, Thermo Scientific Chemicals
CAS: 116422-39-0 Molecular Formula: C4H10O2 Molecular Weight (g/mol): 90.12 MDL Number: MFCD01632593 InChI Key: YTTFFPATQICAQN-BYPYZUCNSA-N Synonym: 2s-2-methoxypropan-1-ol,s-+-2-methoxypropanol,s-2-methoxypropan-1-ol,unii-6725gr28fg,2-methoxy-1-propanol, 2s,1-propanol, 2-methoxy-, 2s,2s-2-methoxy-1-propanol,s-2-methoxy-1-propanol,2-s-methoxypropan-1-ol,1-propanol, 2-methoxy-, s PubChem CID: 642402 IUPAC Name: (2S)-2-methoxypropan-1-ol SMILES: CC(CO)OC