man

Test

CAS RN 117-78-2

OHOOO

CAS RN 117-78-2

IUPAC Name: 9,10-dioxo-9,10-dihydroanthracene-2-carboxylic acid
Synonyms: 9,10-dioxoanthracene-2-carboxylic acid
Molecular Weight (g/mol): 252.23
Molecular Formula: C15H8O4
InChi Key: ASDLSKCKYGVMAI-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C2C(=O)C3=CC=CC=C3C(=O)C2=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results