man

Test

CAS RN 1185165-63-2

CH3SNHNHI

CAS RN 1185165-63-2

IUPAC Name: 4-methylsulfanylcarboximidoylpyridine hydroiodide
Synonyms: 4-methylsulfanylcarboximidoylpyridine hydroiodide
Molecular Weight (g/mol): 280.13
Molecular Formula: C7H9IN2S
InChi Key: CDHPZLSIZALDRX-UHFFFAOYSA-N
SMILES: I.CSC(=N)C1=CC=NC=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results