CAS RN 118628-68-5
CAS RN 118628-68-5
IUPAC Name:
methyl[(1R,2R)-2-(methylazaniumyl)-1,2-diphenylethyl]azanium
Synonyms:
methyl[(1R,2R)-2-(methylammonio)-1,2-diphenylethyl]azanium
Molecular Weight (g/mol):
242.37
Molecular Formula:
C16H22N2
InChi Key:
BTHYVQUNQHWNLS-HZPDHXFCSA-P
SMILES:
C[NH2+][C@@H]([C@H]([NH2+]C)C1=CC=CC=C1)C1=CC=CC=C1