man

Test

CAS RN 118754-53-3

FFFClBrCl

CAS RN 118754-53-3

IUPAC Name: 2-bromo-1,3-dichloro-5-(trifluoromethyl)benzene
Synonyms: 2-bromo-1,3-dichloro-5-(trifluoromethyl)benzene
Molecular Weight (g/mol): 293.89
Molecular Formula: C7H2BrCl2F3
InChi Key: IQHSSYROJYPFDV-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(Cl)=C(Br)C(Cl)=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results