CAS RN 1191-95-3
CAS RN 1191-95-3
IUPAC Name:
cyclobutanone
Synonyms:
cyclobutanone
Molecular Weight (g/mol):
70.09
Molecular Formula:
C4H6O
InChi Key:
SHQSVMDWKBRBGB-UHFFFAOYSA-N
SMILES:
O=C1CCC1
1
–
2
of
2
results
1
Cyclobutanone, 98+%, Thermo Scientific Chemicals
CAS: 1191-95-3 Molecular Formula: C4H6O Molecular Weight (g/mol): 70.09 MDL Number: MFCD00001332 InChI Key: SHQSVMDWKBRBGB-UHFFFAOYSA-N Synonym: unii-6pf2sh405u,cyclobutyloxy,cyclobutanone, 98+%,cyclobutanon,cylcobutanone,cylobutanone,3-cyclobutanone,1-oxocyclobutane,cyclobutanone,pubchem9021 PubChem CID: 14496 IUPAC Name: cyclobutanone SMILES: O=C1CCC1
2
Cyclobutanone, 98%, stab. with ca 0.1% BHT, Thermo Scientific Chemicals
CAS: 1191-95-3 Molecular Formula: C4H6O Molecular Weight (g/mol): 70.09 MDL Number: MFCD00001332 InChI Key: SHQSVMDWKBRBGB-UHFFFAOYSA-N Synonym: unii-6pf2sh405u,cyclobutyloxy,cyclobutanone, 98+%,cyclobutanon,cylcobutanone,cylobutanone,3-cyclobutanone,1-oxocyclobutane,cyclobutanone,pubchem9021 PubChem CID: 14496 IUPAC Name: cyclobutanone SMILES: O=C1CCC1