man

Test

CAS RN 1193-24-4

OHONHN

CAS RN 1193-24-4

IUPAC Name: 6-hydroxy-3,4-dihydropyrimidin-4-one
Synonyms: 6-hydroxy-3H-pyrimidin-4-one
Molecular Weight (g/mol): 112.09
Molecular Formula: C4H4N2O2
InChi Key: DUFGYCAXVIUXIP-UHFFFAOYSA-N
SMILES: OC1=CC(=O)NC=N1
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results