man

Test

CAS RN 1197-55-3

NH2OHO

CAS RN 1197-55-3

IUPAC Name: 2-(4-aminophenyl)acetic acid
Synonyms: benzeneacetic acid, 4-amino-
Molecular Weight (g/mol): 151.17
Molecular Formula: C8H9NO2
InChi Key: CSEWAUGPAQPMDC-UHFFFAOYSA-N
SMILES: NC1=CC=C(CC(O)=O)C=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)