man

Test

CAS RN 119795-57-2

OICl

CAS RN 119795-57-2

IUPAC Name: 1-chloro-4-(3-iodopropoxy)benzene
Synonyms: 1-chloro-4-(3-iodopropoxy)benzene
Molecular Weight (g/mol): 296.53
Molecular Formula: C9H10ClIO
InChi Key: HVVWUBWHLIPSCS-UHFFFAOYSA-N
SMILES: ClC1=CC=C(OCCCI)C=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results