man

Test

CAS RN 120-12-7

CAS RN 120-12-7

IUPAC Name: anthracene
Synonyms: anthracene paranaphthalene anthracene, pure tetra olive n2g tetra olive N2G anthrazen anthracene oil ccris 767 anthracin pure anthracene
Molecular Weight (g/mol): 178.23
Molecular Formula: C14H10
InChi Key: MWPLVEDNUUSJAV-UHFFFAOYSA-N
SMILES: C1=CC2=CC3=CC=CC=C3C=C2C=C1
Quantity 
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
Boiling Point 
  • (6)
  • (5)
Percent Purity 
  • (2)
  • (1)
  • (8)
Physical Form 
  • (2)
  • (5)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (3)
  • (5)
  • (2)
  • (4)
  • (4)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (6)
  • (5)
  • (2)
  • (1)
  • (8)
  • (2)
  • (5)
  • (3)