man

Test

CAS RN 120-71-8

H3COCH3H2N

CAS RN 120-71-8

IUPAC Name: 2-methoxy-5-methylaniline
Synonyms: para-cresidine
Molecular Weight (g/mol): 137.18
Molecular Formula: C8H11NO
InChi Key: WXWCDTXEKCVRRO-UHFFFAOYSA-N
SMILES: COC1=CC=C(C)C=C1N
Molecular Weight (g/mol) 
  • (4)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (4)
Physical Form 
  • (4)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results