CAS RN 12001-85-3
CAS RN 12001-85-3
IUPAC Name:
zinc(2+) bis(3-(3-ethylcyclopentyl)propanoate)
Synonyms:
zinc(2+) bis(3-(3-ethylcyclopentyl)propanoate)
Molecular Weight (g/mol):
403.87
Molecular Formula:
C20H34O4Zn
InChi Key:
HNWHEAHEQDAIDC-UHFFFAOYNA-L
SMILES:
[Zn++].CCC1CCC(CCC([O-])=O)C1.CCC1CCC(CCC([O-])=O)C1
1
–
1
of
1
results
1
Zinc naphthenate, ^=67% in mineral spirits (10% Zn), Thermo Scientific Chemicals
CAS: 12001-85-3 MDL Number: MFCD00147699