CAS RN 12007-16-8
CAS RN 12007-16-8
IUPAC Name:
diboron chromium
Synonyms:
diboron chromium
Molecular Weight (g/mol):
73.62
Molecular Formula:
B2Cr
InChi Key:
UHPOHYZTPBGPKO-UHFFFAOYSA-N
SMILES:
[B].[B].[Cr]
1
–
1
of
1
results
1
Chromium boride, 99% (metals basis), Thermo Scientific Chemicals
CAS: 12007-16-8 Molecular Formula: CrB2 MDL Number: MFCD00016043