CAS RN 12007-23-7
CAS RN 12007-23-7
IUPAC Name:
diboron hafnium
Synonyms:
hafnium diboride
Molecular Weight (g/mol):
200.11
Molecular Formula:
B2Hf
InChi Key:
LRTTZMZPZHBOPO-UHFFFAOYSA-N
SMILES:
[B].[B].[Hf]
1
–
1
of
1
results
1
Hafnium boride, 99.5% (metals basis excluding Zr), Zr <2%, Thermo Scientific Chemicals
CAS: 12007-23-7 Molecular Formula: B2Hf MDL Number: MFCD00016125