CAS RN 12007-62-4
CAS RN 12007-62-4
IUPAC Name:
magnesium(2+) boron oxidandiide
Synonyms:
magnesium(2+) boron oxidandiide
Molecular Weight (g/mol):
51.11
Molecular Formula:
BMgO
InChi Key:
YZUWTDQUEUJLAR-UHFFFAOYSA-N
SMILES:
[B].[O--].[Mg++]
1
–
1
of
1
results
1
Magnesium tetraborate, 99%, Thermo Scientific Chemicals
CAS: 12007-62-4 Molecular Formula: B2Mg3O6 Molecular Weight (g/mol): 190.529 MDL Number: MFCD02688001 InChI Key: NFMWFGXCDDYTEG-UHFFFAOYSA-N Synonym: acmc-1bvay,boric acid h3bo3 , magnesium salt 2:3,ksc171o6l,trimagnesium 2+ diborate,parent,trimagnesium 2+ ion diborate PubChem CID: 21981398 IUPAC Name: trimagnesium;diborate SMILES: B([O-])([O-])[O-].B([O-])([O-])[O-].[Mg+2].[Mg+2].[Mg+2]