man

Test

CAS RN 1202-39-7

HOOClCl

CAS RN 1202-39-7

IUPAC Name: (2E)-3-(3,4-dichlorophenyl)prop-2-enoic acid
Synonyms: (2E)-3-(3,4-dichlorophenyl)prop-2-enoic acid
Molecular Weight (g/mol): 217.05
Molecular Formula: C9H6Cl2O2
InChi Key: RRLUFPHCTSFKNR-DUXPYHPUSA-N
SMILES: OC(=O)\C=C\C1=CC=C(Cl)C(Cl)=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)