man

Test

CAS RN 12021-95-3

ZrFFFFFF

CAS RN 12021-95-3

IUPAC Name: hexafluorodihydridozirconium
Synonyms: hexafluorodihydridozirconium
Molecular Weight (g/mol): 207.23
Molecular Formula: F6H2Zr
InChi Key: PZFQYBOZXGWNAM-UHFFFAOYSA-H
SMILES: F[ZrH2](F)(F)(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)