man

Test

CAS RN 12030-24-9

InInSSS

CAS RN 12030-24-9

IUPAC Name: indium sulfane
Synonyms: indium(III) sulfide
Molecular Weight (g/mol): 148.89
Molecular Formula: H2InS
InChi Key: GXBLCDVIPOLQNI-UHFFFAOYSA-N
SMILES: S.[In]
Percent Purity 
  • (3)
  • (3)
  • (3)
Quantity 
  • (3)
  • (3)
  • (3)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (3)
  • (9)
  • (3)
  • (3)
  • (3)