man

Test

CAS RN 12035-38-0

NiSnO3

CAS RN 12035-38-0

IUPAC Name: tin(4+) λ³-nickel(3+) trioxidandiide
Synonyms: tin(4+) λ³-nickel(3+) trioxidandiide
Molecular Weight (g/mol): 225.4
Molecular Formula: NiO3Sn
InChi Key: SHULAURIUQUJKN-UHFFFAOYSA-N
SMILES: [O--].[O--].[O--].[Ni+3].[Sn+4]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results