man

Test

CAS RN 12037-42-2

OVOOVOOVOOVOOVOOVOO

CAS RN 12037-42-2

IUPAC Name: [(dioxovanadio)oxy]dioxovanadium; tetrakis(dioxovanadium)
Synonyms: tetrakis(vanadium dioxide); vanadium(V) oxide
Molecular Weight (g/mol): 513.64
Molecular Formula: O13V6
InChi Key: FTGZPVKRMCXHDZ-UHFFFAOYSA-N
SMILES: O=[V]=O.O=[V]=O.O=[V]=O.O=[V]=O.O=[V](=O)O[V](=O)=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results