man

Test

CAS RN 12038-21-0

SPtS

CAS RN 12038-21-0

IUPAC Name: disulfanylideneplatinum
Synonyms: disulfanylideneplatinum
Molecular Weight (g/mol): 259.2
Molecular Formula: PtS2
InChi Key: RKNFJIIYAUSTJA-UHFFFAOYSA-N
SMILES: S=[Pt]=S

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results