man

Test

CAS RN 12057-17-9

LiMnOMn4

CAS RN 12057-17-9

IUPAC Name: manganese(4+) manganese(3+) lithium(1+) tetraoxidandiide
Synonyms: manganese(4+) manganese(3+) lithium(1+) tetraoxidandiide
Molecular Weight (g/mol): 180.81
Molecular Formula: LiMn2O4
InChi Key: KEZPOJYZIWMPGY-UHFFFAOYSA-N
SMILES: [Li+].[O--].[O--].[O--].[O--].[Mn+3].[Mn+4]

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results