man

Test

CAS RN 12060-08-1

OOOScSc

CAS RN 12060-08-1

IUPAC Name: discandium(3+) trioxidandiide
Synonyms: scandium oxide
Molecular Weight (g/mol): 137.91
Molecular Formula: O3Sc2
InChi Key: HJGMWXTVGKLUAQ-UHFFFAOYSA-N
SMILES: [O--].[O--].[O--].[Sc+3].[Sc+3]
Quantity 
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (4)
  • (2)
Physical Form 
  • (10)
Grade 
  • (5)
  • (2)
For Use With (Equipment) 
  • (6)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (4)
  • (2)
  • (10)
  • (5)
  • (2)
  • (6)
  • (2)