man

Test

CAS RN 12063-19-3

HOFeOHOFeOZn

CAS RN 12063-19-3

IUPAC Name: bis(ironoylol) zinc
Synonyms: bis(iron hydroxide) zinc
Molecular Weight (g/mol): 243.08
Molecular Formula: Fe2H2O4Zn
InChi Key: NNGHIEIYUJKFQS-UHFFFAOYSA-L
SMILES: [Zn].O[Fe]=O.O[Fe]=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results