CAS RN 12070-12-1
CAS RN 12070-12-1
IUPAC Name:
tungsteniomethanide
Synonyms:
tungsten carbide
Molecular Weight (g/mol):
195.85
Molecular Formula:
CW
InChi Key:
OAXLZNWUNMCZSO-UHFFFAOYSA-N
SMILES:
[C-]#[W]
1
–
2
of
2
results
1
Tungsten carbide, 99% (metals basis), Thermo Scientific Chemicals
CAS: 12070-12-1 Molecular Formula: CW Molecular Weight (g/mol): 195.85 MDL Number: MFCD00011464 InChI Key: UONOETXJSWQNOL-UHFFFAOYSA-N Synonym: tungsten carbide,tungsten iv carbide,rfdbds g@,tungsten carbide ds 60,methanidylidynetungstenylium,methanidylidynetungsten 1+,tungsten-carbide/cobalt co=8wt% powder nano IUPAC Name: tungsteniomethanide SMILES: [C-]#[W+]
2
Tungsten carbide, 99.5% (metals basis), Thermo Scientific Chemicals
CAS: 12070-12-1 Molecular Formula: CW Molecular Weight (g/mol): 195.85 MDL Number: MFCD00011464 InChI Key: UONOETXJSWQNOL-UHFFFAOYSA-N Synonym: tungsten carbide,tungsten iv carbide,rfdbds g@,tungsten carbide ds 60,methanidylidynetungstenylium,methanidylidynetungsten 1+,tungsten-carbide/cobalt co=8wt% powder nano IUPAC Name: methanidylidynetungstenylium SMILES: [C-]#[W+]