man

Test

CAS RN 120743-99-9

H3CH3CCH3SiH3CCH3OO

CAS RN 120743-99-9

IUPAC Name: 4-[(tert-butyldimethylsilyl)oxy]benzaldehyde
Synonyms: 4-[(tert-butyldimethylsilyl)oxy]benzaldehyde
Molecular Weight (g/mol): 236.39
Molecular Formula: C13H20O2Si
InChi Key: XACWSBWCLJXKGI-UHFFFAOYSA-N
SMILES: CC(C)(C)[Si](C)(C)OC1=CC=C(C=O)C=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results