CAS RN 12078-25-0
CAS RN 12078-25-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Dicarbonylcyclopentadienylcobalt, Thermo Scientific Chemicals
CAS: 12078-25-0 MDL Number: MFCD00009589