man

Test

CAS RN 1215194-05-0

CH3HONHCl

CAS RN 1215194-05-0

IUPAC Name: 1-phenyl-2-(pyrrolidin-1-yl)propan-1-ol hydrochloride
Synonyms: 1-phenyl-2-(pyrrolidin-1-yl)propan-1-ol hydrochloride
Molecular Weight (g/mol): 241.76
Molecular Formula: C13H20ClNO
InChi Key: PMKBLGNPWCWAAH-UHFFFAOYNA-N
SMILES: Cl.CC(C(O)C1=CC=CC=C1)N1CCCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results