CAS RN 12159-07-8
CAS RN 12159-07-8
IUPAC Name:
pentacopper silicon
Synonyms:
copper silicide
Molecular Weight (g/mol):
345.82
Molecular Formula:
Cu5Si
InChi Key:
JUZTWRXHHZRLED-UHFFFAOYSA-N
SMILES:
[Si].[Cu].[Cu].[Cu].[Cu].[Cu]
1
–
1
of
1
results
1
Copper silicide, 99.5% (metals basis), Thermo Scientific Chemicals
CAS: 12159-07-8 Molecular Formula: Cu5Si MDL Number: MFCD00134719