man

Test

CAS RN 122433-41-4

CH3ONOOH3CBr

CAS RN 122433-41-4

IUPAC Name: methyl 5-bromo-2-methoxypyridine-3-carboxylate
Synonyms: methyl 5-bromo-2-methoxypyridine-3-carboxylate
Molecular Weight (g/mol): 246.06
Molecular Formula: C8H8BrNO3
InChi Key: OLQOVEOEHQDKSC-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(Br)=CN=C1OC

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results