man

Test

CAS RN 122957-50-0

CH3NNN

CAS RN 122957-50-0

IUPAC Name: 4-(2-methyl-1H-imidazol-1-yl)benzonitrile
Synonyms: 4-(2-methylimidazol-1-yl)benzonitrile
Molecular Weight (g/mol): 183.21
Molecular Formula: C11H9N3
InChi Key: FAMJRKMLLLJOGZ-UHFFFAOYSA-N
SMILES: CC1=NC=CN1C1=CC=C(C=C1)C#N

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results