man

Test

CAS RN 123-06-8

CH3ONN

CAS RN 123-06-8

IUPAC Name: 2-(ethoxymethylidene)propanedinitrile
Synonyms: 2-(ethoxymethylidene)propanedinitrile
Molecular Weight (g/mol): 122.13
Molecular Formula: C6H6N2O
InChi Key: OEICGMPRFOJHKO-UHFFFAOYSA-N
SMILES: CCOC=C(C#N)C#N
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
Boiling Point 
  • (3)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results