man

Test

CAS RN 123-75-1

HClHN

CAS RN 123-75-1

IUPAC Name: hydrogen pyrrolidine chloride
Synonyms: pyrrolidine
Molecular Weight (g/mol): 107.58
Molecular Formula: C4H10ClN
InChi Key: FCLZCOCSZQNREK-UHFFFAOYSA-N
SMILES: [H+].[Cl-].C1CCNC1
Quantity 
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)
  • (2)
  • (7)
Percent Purity 
  • (3)
  • (5)
  • (2)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (1)
  • (7)
  • (3)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (7)
  • (3)
  • (5)
  • (2)
  • (4)