CAS RN 123333-87-9
CAS RN 123333-87-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Rhodium(III) bromide hydrate, Thermo Scientific Chemicals
CAS: 123333-87-9 Molecular Formula: Br3Rh Molecular Weight (g/mol): 342.62 MDL Number: MFCD00149837 InChI Key: MMRXYMKDBFSWJR-UHFFFAOYSA-K IUPAC Name: rhodium(3+) tribromide SMILES: [Br-].[Br-].[Br-].[Rh+3]