man

Test

CAS RN 123572-63-4

FH2NOFFF

CAS RN 123572-63-4

IUPAC Name: 5-fluoro-2-(trifluoromethoxy)aniline
Synonyms: 5-fluoro-2-(trifluoromethoxy)aniline
Molecular Weight (g/mol): 195.12
Molecular Formula: C7H5F4NO
InChi Key: AUJAYNLPYRARLU-UHFFFAOYSA-N
SMILES: NC1=C(OC(F)(F)F)C=CC(F)=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results