man

Test

CAS RN 1240256-85-2

SFFFFFFOCl

CAS RN 1240256-85-2

IUPAC Name: 2-fluoro-4-(pentafluoro-λ⁶-sulfanyl)benzoyl chloride
Synonyms: 2-fluoro-4-(pentafluoro-λ⁶-sulfanyl)benzoyl chloride
Molecular Weight (g/mol): 284.6
Molecular Formula: C7H3ClF6OS
InChi Key: ZJOFYWIDVCEMTP-UHFFFAOYSA-N
SMILES: FC1=C(C=CC(=C1)S(F)(F)(F)(F)F)C(Cl)=O

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results