man

Test

CAS RN 125700-69-8

BFFFFH3CNCH3NH3CCH3ONONN

CAS RN 125700-69-8

IUPAC Name: [(dimethylamino)[(4-oxo-3,4-dihydro-1,2,3-benzotriazin-3-yl)oxy]methylidene]dimethylazanium; tetrafluoroboranuide
Synonyms: [(dimethylamino)[(4-oxo-1,2,3-benzotriazin-3-yl)oxy]methylidene]dimethylazanium tetrafluoroborate
Molecular Weight (g/mol): 349.1
Molecular Formula: C12H16BF4N5O2
InChi Key: FOBCPCIJLQTYBT-UHFFFAOYSA-N
SMILES: F[B-](F)(F)F.CN(C)C(ON1N=NC2=CC=CC=C2C1=O)=[N+](C)C
Quantity 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)