CAS RN 1261673-70-4
CAS RN 1261673-70-4
IUPAC Name:
2-chloro-6-(trifluoromethoxy)aniline
Synonyms:
2-chloro-6-(trifluoromethoxy)aniline
Molecular Weight (g/mol):
211.57
Molecular Formula:
C7H5ClF3NO
InChi Key:
NNDFBODLWFJOSA-UHFFFAOYSA-N
SMILES:
NC1=C(OC(F)(F)F)C=CC=C1Cl
1
–
1
of
1
results
1
2-Chloro-6-(trifluoromethoxy)aniline, 97%, Thermo Scientific™
CAS: 1261673-70-4 Molecular Formula: C7H5ClF3NO Molecular Weight (g/mol): 211.568 MDL Number: MFCD07779302 InChI Key: NNDFBODLWFJOSA-UHFFFAOYSA-N Synonym: 2-chloro-6-trifluoromethoxy aniline PubChem CID: 86277618 IUPAC Name: 2-chloro-6-(trifluoromethoxy)aniline SMILES: C1=CC(=C(C(=C1)Cl)N)OC(F)(F)F